Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium acetate 99.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid barium salt Barium di(acetate)

012198-30 012198-A1 12198-A7 12198-30 12198-A1 012198-A7
AA12198-A7EA 622.99 USD InStock
CAAA12198-A7 AA12198-A7 CAAA12198-30 AA12198-30 CAAA12198-A1 AA12198-A1
Barium acetate 99.0-102.0% ACS
Barium acetate


Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Melting Pt: ∼450 °C
Density: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 13,00968
Order Now
Specification Test Results
Insoluble matter 0.01%
Cl 0.1%
Oxidizing substances (as NO3) 0.005%
K 0.003%
Na 0.005%
Ca 0.05%
Heavy Metals (as Pb) 5 ppm max.
Fe 0.001%
Sr Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty