Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium(II) nitrate tetrahydrate ≥98.5%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cadmium nitrate tetrahydrate

12228-36 12228-22 012228-36 012228-22
AA12228-22EA 47.97 USD InStock
AA12228-22 CAAA12228-36 CAAA12228-22 AA12228-36
Cadmium(II) nitrate tetrahydrate ≥98.5%
Cadmium (II) nitrate tetrahydrate
Manufacturing of Cd salts, photographic emulsions, laboratory reagent, coloring glass


Formula: CdN₂O₆·4H₂O
MW: 308.48 g/mol
Boiling Pt: 132 °C
Melting Pt: 59 °C
Density: 2,45
Storage Temperature: Ambient
MDL Number: MFCD00149626
CAS Number: 10022-68-1
EINECS: 233-710-6
UN: 3087
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty