Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gallium phosphide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Gallium phosphide

12266-09 012266-09 12266-18 12266-04 012266-04 012266-18
AA12266-18EA 2145.99 USD InStock
CAAA12266-04 CAAA12266-09 AA12266-18 CAAA12266-18 AA12266-04 AA12266-09
Gallium phosphide ≥99.999% (metals basis)
Gallium phosphide
In semiconducting devices


Formula: GaP
MW: 100.7 g/mol
Melting Pt: 1465 °C
Density: 4.13
Storage Temperature: Ambient
MDL Number: MFCD00016109
CAS Number: 12063-98-8
EINECS: 235-057-2
Merck Index: 12,04366
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty