Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) acetate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid mercury (II) salt Mercuric acetate Mercury diacetate

12273-30 012273-30 012273-18 12273-18
AA12273-30EA 214.99 USD InStock
CAAA12273-18 AA12273-30 CAAA12273-30 AA12273-18
Mercury(II) acetate ≥98.0% ACS
Mercury (II) acetate


Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 179…182 °C
Density: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898
Order Now
Specification Test Results
Insoluble matter 0.01%
Residue after reduction 0.02%
Cl 0.005%
NO3 0.005%
SO3 0.005%
Heavy Metals (as Pb) 0.002%
Fe 0.001%
Mercurous mercury (as Hg) 0.4%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty