Synonyms:
Adam's type catalyst Platinum dioxide monohydrate
12278-03
12278-06
012278-06
012278-03
AA12278-03EA
240
USD
InStock
AA12278-03
CAAA12278-06
CAAA12278-03
AA12278-06
Platinium(IV) oxide monohydrate Pt ≥ 75% (≥75% Pt)
Platinium (IV) oxide monohydrate
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups