Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinium(IV) oxide monohydrate Pt ≥ 75% (≥75% Pt)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Adam's type catalyst Platinum dioxide monohydrate

12278-03 12278-06 012278-06 012278-03
AA12278-03EA 240 USD InStock
AA12278-03 CAAA12278-06 CAAA12278-03 AA12278-06
Platinium(IV) oxide monohydrate Pt ≥ 75% (≥75% Pt)
Platinium (IV) oxide monohydrate
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups


Formula: O₂Pt·1H₂O
MW: 245.09 g/mol
Melting Pt: 450 °C
Storage Temperature: Ambient
MDL Number: MFCD00066964
CAS Number: 12137-21-2
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty