Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tellurium tetrachloride ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tellurium(IV) chloride

012284-14 012284-22 12284-22 12284-14
AA12284-14EA 129.99 USD InStock
AA12284-14 AA12284-22 CAAA12284-14 CAAA12284-22
Tellurium tetrachloride ≥99.9% (metals basis)
Tellurium tetrachloride


Formula: Cl₄Te
MW: 269.41 g/mol
Boiling Pt: 380 °C
Melting Pt: 224 °C
Density: 3,01
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS Number: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8,II
Merck Index: 12,09279
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty