Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tetra-Chloroauric(III) acid hydrate (≥49% Au) ≥99.9% (metals basis) Au ≥ 49.0%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen tetrachloroaurate (III) hydrate tetra-Chloroauric acid hydrate

012325-14 12325-14 12325-06 012325-03 012325-06 12325-03
AA12325-06EA 792.99 USD InStock
AA12325-06 CAAA12325-06 AA12325-14 AA12325-03 CAAA12325-14 CAAA12325-03
tetra-Chloroauric(III) acid hydrate (≥49% Au) ≥99.9% (metals basis) Au ≥ 49.0%
tetra-Chloroauric (III) acid hydrate
Crystals


Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
Density: 1.169 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty