Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium nitrate nonahydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aluminum nitrate nonahydrate

12360-A1 012360-14 012360-30 12360-A7 12360-30 12360-14 012360-A7 012360-A1
AA12360-30EA 87.22 USD InStock
CAAA12360-30 CAAA12360-A1 AA12360-30 CAAA12360-A7 AA12360-14 AA12360-A7 AA12360-A1 CAAA12360-14
Aluminium nitrate nonahydrate ≥98%
Aluminium nitrate nonahydrate


Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73.5 °C
Density: 1,72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003419
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty