Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium chloride ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ammonium muriateSalmiac

012361-A3 12361-36 012361-36 12361-A3
AA12361-A3EA 111.75 USD InStock
AA12361-A3 CAAA12361-36 AA12361-36 CAAA12361-A3
Ammonium chloride ≥99.5%
Ammonium chloride
As a flux in galvanizing steel sheet, in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton


Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 340 °C (subl.)
Density: 1.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty