Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2,4-Trimethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pseudocumene

12411-A3 012411-36 12411-A9 12411-36 012411-A3 012411-A9 12411-B4 012411-B4
AA12411-A3EA 210.99 USD InStock
AA12411-A3 AA12411-B4 AA12411-36 CAAA12411-A3 CAAA12411-B4 CAAA12411-36 CAAA12411-A9 AA12411-A9
1,2,4-Trimethylbenzene 98%
1,2,4-Trimethylbenzene


Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 169…171 °C
Melting Pt: –44 °C
Density: 0.882
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 3295
ADR: 3,III
Merck Index: 12,08100
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty