Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(II) chloride hydrate ≥99%, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

012484-14 012484-30 12484-A7 12484-30 012484-A1 12484-A1 12484-14 012484-A7
AA12484-14EA 50.19 USD InStock
AA12484-14 CAAA12484-A7 AA12484-A7 CAAA12484-30 AA12484-30 CAAA12484-A1 AA12484-A1 CAAA12484-14
Iron(II) chloride hydrate ≥99%, Reagent Grade
Iron(II) chloride hydrate
Applications: Reducing agent in metallurgy, in pharmaceutical preparations, as mordant in dyeing and in sewage treatment


Formula: Cl₂Fe•xH₂O
MW: 126.745 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149711
CAS Number: 23838-02-0
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty