Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium nickel (II) sulfate hexahydrate

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel ammonium sulfate hexahydrate

012519-36 12519-36 12519-22 012519-22
AA12519-36EA 384.99 USD InStock
AA12519-36 AA12519-22 CAAA12519-36 CAAA12519-22
Ammonium nickel (II) sulfate hexahydrate
Ammonium nickel (II) sulphate hexahydrate


Formula: H₈N₂NiO₈S₂·6H₂O
MW: 394.99 g/mol
Density: 1,923
Storage Temperature: Ambient
MDL Number: MFCD00150260
CAS Number: 7785-20-8
EINECS: 239-793-5
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty