Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Copper dinitrate hemipentahydrate Cupric nitrate hemipentahydrate

12523-18 12523-36 012523-36 012523-18 12523-A3 012523-A9 012523-A3 12523-A9
AA12523-A9EA 1099.99 USD InStock
AA12523-A9 CAAA12523-A3 CAAA12523-36 AA12523-A3 AA12523-36 CAAA12523-18 CAAA12523-A9 AA12523-18
Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS
Copper (II) nitrate hemipentahydrate
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black, and producing a burnished effect on iron


Formula: Cu(NO₃)₂·2,5H₂O
MW: 232.59 g/mol
Boiling Pt: 170 °C (dec.)
Melting Pt: ∼115 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD00149671
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III
Merck Index: 13,02671
Order Now
Specification Test Results
Insoluble matter 0.01%
Cl 0.002%
SO3 0.01%
Ca 0.005%
Ni 0.01%
K 0.005%
Na 0.0001
Pb 0.00001
Fe 0.00005

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty