Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium(III) acetylacetonate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III) Chromium (III) 2,4-pentanedionate Chromium (III) tris(2,4-pentanedionato-O,O')

12538-22 12538-36 012538-22 012538-36
AA12538-22EA 74.44 USD InStock
CAAA12538-22 AA12538-22 CAAA12538-36 AA12538-36
Chromium(III) acetylacetonate 97%
Chromium (III) acetylacetonate
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD


Formula: C₁₅H₂₁CrO₆
MW: 349.32 g/mol
Boiling Pt: 340 °C
Melting Pt: 216 °C
Density: 1,35
Storage Temperature: Ambient
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty