Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Selenious acid 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Selenous acid

12574-30 12574-30 012574-06 012574-18 012574-30 12574-06
AA12574-30AEA 242.99 USD InStock
CAAA12574-06A CAAA12574-18A AA12574-30A CAAA12574-30A AA12574-06A AA12574-18A
Selenious acid 97%
Selenious acid
As an oxidizing agent, as a reagent for alkaloids


Formula: H₂O₃Se
MW: 128.97 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3290
ADR: 6.1,II
Merck Index: 14,08430
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty