Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Lithium tetraborate ≥99.6% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Lithium tetraborate

012592-30 12592-18 012592-18 12592-A1 12592-30 12592-A1
AA12592-A1EA 761.19 USD InStock
AA12592-A1 AA12592-18 CAAA12592-18 CAAA12592-30 CAAA12592-A1 AA12592-30
di-Lithium tetraborate ≥99.6% (metals basis)
di-Lithium tetraborate


Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 930 °C
Density: 2.4
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty