Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenate, anhydrous ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12613-18 012613-A1 12613-A1 012613-06 12613-06 012613-30 12613-30 012613-18
AA12613-A1EA 531.99 USD InStock
AA12613-A1 CAAA12613-A1 AA12613-30 CAAA12613-30 AA12613-18 AA12613-06 CAAA12613-06 CAAA12613-18
Sodium selenate, anhydrous ≥99.8% (metals basis)
Sodium selenate
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.

Stable under alkaline and oxidizing conditions.


Formula: Na₂O₄Se
MW: 188.94 g/mol
Melting Pt: 32 °C trans.
Density: 3.213
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS Number: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Merck Index: 12,08820
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty