Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium sulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12839-18 012839-09 12839-09 12839-04 12839-04 012839-30 012839-18
AA12839-30EA 1907.99 USD InStock
AA12839-30 AA12839-09 CAAA12839-09 AA12839-18 CAAA12839-18 AA12839-04 CAAA12839-04
Lithium sulfide ≥99.9% (metals basis)
Lithium sulphide
  • -200 Mesh Powder


Formula: Li₂S
MW: 45.95 g/mol
Melting Pt: 900…975 °C
Density: 1.66
Storage Temperature: Ambient
MDL Number: MFCD00011085
CAS Number: 12136-58-2
EINECS: 235-228-1
UN: 3134
ADR: 4.3,II
Order Now
Specification Test Results
Assay Minimum 99.9

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty