Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium sulfide ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Barium(II) sulfideBarium(II) sulphide

012848-09 12848-09 12848-18 012848-18
AA12848-18EA 423.86 USD InStock
CAAA12848-09 AA12848-18 AA12848-09 CAAA12848-18
Barium sulfide ≥99.7% (metals basis)
Barium sulphide


Formula: BaS
MW: 169.4 g/mol
Melting Pt: 1200 °C
Density: 4,25
Storage Temperature: Ambient
MDL Number: MFCD00014192
CAS Number: 21109-95-5
EINECS: 244-214-4
UN: 3134
ADR: 4.3,III
Merck Index: 12,01024
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty