Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Praseodymium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Praseodymium (III) nitrate

12909-A1 012909-30 012909-18 12909-A1 12909-09 12909-30 12909-18 012909-09
AA12909-30EA 308.99 USD InStock
CAAA12909-A1 AA12909-30 AA12909-A1 CAAA12909-18 AA12909-18 CAAA12909-09 CAAA12909-30 AA12909-09
Praseodymium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Praseodymium(III) nitrate hydrate
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide


Formula: N₃O₉Pr
MW: 326.92 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011175
CAS Number: 10361-80-5
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty