Would you like to log in again?
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Supplier: Thermo Scientific Chemicals
12942-09 012942-03 012942-09 12942-03
AA12942-03EA 88.92 USD InStock
CAAA12942-09 AA12942-03 CAAA12942-03 AA12942-09
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
Expiration date provided
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty