Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc fluoride ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Zinc (II) fluoride

012986-06 12986-22 12986-22 012986-14 12986-14 12986-06
AA12986-14EA 261.99 USD InStock
CAAA12986-14 CAAA12986-06 AA12986-14 AA12986-06 CAAA12986-22 AA12986-22
Zinc fluoride ≥99.995% (metals basis), Puratronic®
Zinc fluoride


Formula: F₂Zn
MW: 103.39 g/mol
Boiling Pt: 1500 °C
Melting Pt: 872 °C
Density: 4.95
Storage Temperature: Ambient
MDL Number: MFCD00011298
CAS Number: 7783-49-5
EINECS: 232-001-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,10136
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty