Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium oxalate monohydrate ≥99.9985% (metals basis) (excluding 25 ppm max alkali earths), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Calcium ethanedioate monohydrate Oxalic acid calcium salt monohydrate

013007-14 013007-06 13007-14 13007-06
AA13007-06EA 58.39 USD InStock
AA13007-06 CAAA13007-14 AA13007-14 CAAA13007-06
Calcium oxalate monohydrate ≥99.9985% (metals basis) (excluding 25 ppm max alkali earths), Puratronic®
Calcium oxalate monohydrate


Formula: C₂CaO₄·1H₂O
MW: 146.11 g/mol
Melting Pt: 200 °C
Density: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00150039
CAS Number: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty