Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium wolframate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Calcium tungstateCalcium tungsten oxide

13051-36 13051.36 13051-14 13051-22 13051.14 13051.22
AA13051-36EA 316.99 USD InStock
AA13051-36 AA13051-14 CAAA13051-36 CAAA13051-14 AA13051-22 CAAA13051-22
Calcium wolframate 98%
Calcium wolframate
Insoluble in H2O. Decomposes by hot HCl, HNO3


Formula: CaO₄W
MW: 287.92 g/mol
Melting Pt: 1620 °C
Storage Temperature: Ambient
MDL Number: MFCD00010913
CAS Number: 7790-75-2
EINECS: 232-219-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty