Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecacarbonyltriiron ≥96% (dry weight) stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triiron dodecacarbonyl

Stabilization: Stabilized with methanol 5-10%

13059-04 13059.09 13059.18 013059-04 13059-18 13059-09
AA13059-04EA 31.29 USD InStock
AA13059-04 CAAA13059-04 AA13059-18 CAAA13059-09 CAAA13059-18 AA13059-09
Dodecacarbonyltriiron ≥96% (dry weight) stabilized
Dodecacarbonyltriiron
Reducing agent (reduces NO2), catalyst for alkyne trimerization.


Formula: C₁₂Fe₃O₁₂
MW: 503.66 g/mol
Melting Pt: 140 °C (dec.)
Density: 2.00
Storage Temperature: Ambient
MDL Number: MFCD00135617
CAS Number: 17685-52-8
EINECS: 241-668-5
UN: 3179
ADR: 4.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty