Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) fluoride, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel difluoride

013067-18 013067-09 13067-09 13067-18
AA13067-18EA 264.99 USD InStock
CAAA13067-18 AA13067-18 CAAA13067-09 AA13067-09
Nickel(II) fluoride, anhydrous 97%
Nickel (II) fluoride


Formula: F₂Ni
MW: 96.69 g/mol
Melting Pt: 1360…1380 °C
Density: 4.72
Storage Temperature: Ambient
MDL Number: MFCD00016263
CAS Number: 10028-18-9
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III
Merck Index: 12,06591
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty