Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentatitanium trisilicide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Titanium silicide

13087-18 13087-30 13087.3 13087.18
AA13087-18EA 139.99 USD InStock
CAAA13087-18 AA13087-18 AA13087-30 CAAA13087-30
Pentatitanium trisilicide ≥99.5% (metals basis)
Pentatitanium trisilicide


Formula: Si₃Ti₅
Melting Pt: 2130 °C
Storage Temperature: Ambient
MDL Number: MFCD01310208
CAS Number: 12067-57-1
EINECS: 235-082-9
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty