Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt sulfide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cobalt (II) sulphide

13114-22 13114-14 013114-14 013114-22
AA13114-14EA 103.99 USD InStock
CAAA13114-14 AA13114-14 CAAA13114-22 AA13114-22
Cobalt sulfide ≥99.5% (metals basis)
Cobalt sulphide


Formula: CoS
MW: 91 g/mol
Melting Pt: >1116 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016032
CAS Number: 1317-42-6
EINECS: 215-273-3
UN: 3077
ADR: 9,III
Merck Index: 12,02511
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty