Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bismuth (III) selenide

13126-18 13126-09 013126-18 013126-09
AA13126-18EA 365.99 USD InStock
CAAA13126-09 AA13126-18 AA13126-09 CAAA13126-18
Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition
Dibismuth triselenide
In semiconductor research, multilayer thin-film, magnetoresistant film


Formula: Bi₂Se₃
MW: 654.84 g/mol
Melting Pt: 710 °C
Density: 6.82
Storage Temperature: Ambient
MDL Number: MFCD00014200
CAS Number: 12068-69-8
EINECS: 235-104-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,01319
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty