Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diantimony triselenide ≥99.999% (metals basis), pieces

Supplier: Thermo Scientific Chemicals
Danger

013130-06 013130-14 13130-06 13130-14
AA13130-06EA 151.99 USD InStock
AA13130-06 CAAA13130-06 AA13130-14 CAAA13130-14
Diantimony triselenide ≥99.999% (metals basis), pieces
Diantimony triselenide


Formula: Sb₂Se₃
MW: 480.4 g/mol
Melting Pt: 611 °C
Density: 5.81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016319
CAS Number: 1315-05-5
EINECS: 215-256-0
UN: 3077
ADR: 9,III
Merck Index: 12,00753
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty