Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium nitrate, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Danger

013405.A7 013405-A1 013405-30 013405-14
AA13405-A1EA 438.71 USD InStock
AA13405-A1 AA13405-30 AA13405-A7 AA13405-14
Lithium nitrate, anhydrous 99%
Lithium nitrate
Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant


Formula: LiNO₃
MW: 68.95 g/mol
Boiling Pt: 600 °C (dec.)
Melting Pt: 264 °C
Density: 2.38
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 13,05558
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty