Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chromate tetrahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

013453-A1 13453-30 13453-A1 013453-30
AA13453-A1EA 259.08 USD InStock
CAAA13453-A1 AA13453-A1 CAAA13453-30 AA13453-30
Sodium chromate tetrahydrate, REacton®, Reagent Grade
Sodium chromate tetrahydrate
Corrosion inhibitor, pigment manufacture


Formula: CrNa₂O₄·4H₂O
MW: 234.03 g/mol
Melting Pt: 792.2 °C
Density: 2.73
Storage Temperature: Ambient
MDL Number: MFCD00149167
CAS Number: 10034-82-9
EINECS: 231-889-5
UN: 3288
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty