Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, anhydrous ≥97.0% ACS low chloride

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

013455-F6 13455-22 13455-A9 013455-A3 13455-A3 13455-F6 013455-22 013455-A9 013455-36 13455-36
AA13455-A3EA 114.82 USD InStock
CAAA13455-36 AA13455-A3 AA13455-36 CAAA13455-22 CAAA13455-F6 AA13455-F6 CAAA13455-A3 AA13455-22 CAAA13455-A9 AA13455-A9
Sodium hydroxide, anhydrous ≥97.0% ACS low chloride
Sodium hydroxide
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Cl 0.005%
N 0.1%
PO9 0.001%
SO3 0.003%
Heavy Metals (as Ag) 0.002%
Fe 0.001%
Hg 0.1 ppm
Ca 0.005%
Mg 0.002%
Ni 0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty