Would you like to log in again?
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Supplier: Thermo Scientific Chemicals
13478-CH 013478-RU 13478-CH 013478-RR 13478-RU 13478-RR
AA13478-RREA 50.89 USD InStock
AA13478-RR CAAA13478-RU CAAA13478-RR AA13478-CH CAAA13478-CH AA13478-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Stainless steel
Open area: 36%; Width of opening: 0.015in
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty