Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium chloride, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13686-14 013686-14
AA13686-14EA 193 USD InStock
CAAA13686-14 AA13686-14
Barium chloride, ultra dry ≥99.998% (metals basis)
Barium chloride


Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560 °C
Melting Pt: 963 °C
Density: 3.9
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty