Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) formate dihydrate

Supplier: Thermo Scientific Chemicals
Danger

13801.36 13801-A3 13801-22 13801-36 13801.22 013801.A3
AA13801-A3EA 663.99 USD InStock
CAAA13801-36 AA13801-A3 AA13801-36 CAAA13801-A3 CAAA13801-22 AA13801-22
Nickel(II) formate dihydrate
Nickel(II) formate dihydrate
Applications: Nickel catalyst preparations
In the range 180-200° decomposes to Ni, CO, CO2, H2, H2O, CH3. Partially soluble in water


Formula: Ni(HCO₂)₂.2H₂O
MW: 184.77 g/mol (148.74 anhy)
Melting Pt: 130…140 °C (-2H₂O)
Density: 2,15
Storage Temperature: Ambient
MDL Number: MFCD00153034
CAS Number: 15694-70-9
EINECS: 222-101-0
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty