Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium ammonium sulfate dodecahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Aluminium ammonium bis(sulphate) dodecahydrate

13802-A3 13802-22 13802-36 013802-36 013802-A3 013802-22
AA13802-36EA 86.79 USD InStock
AA13802-36 CAAA13802-22 CAAA13802-A3 AA13802-22 CAAA13802-36 AA13802-A3
Aluminium ammonium sulfate dodecahydrate, REacton®, Reagent Grade
Aluminium ammonium sulphate dodecahydrate
Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper plating


Formula: AlNH₄(SO₄)₂·12H₂O
MW: 453.33 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 93…94 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00051042
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty