Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indium phosphide ≥99.9999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Indium phosphide

013974-03 013974-06 13974-06 13974-03
AA13974-03EA 218.99 USD InStock
CAAA13974-03 AA13974-03 CAAA13974-06 AA13974-06
Indium phosphide ≥99.9999% (metals basis)
Indium phosphide


Formula: InP
MW: 145.79 g/mol
Melting Pt: 1070 °C
Density: 4.81
Storage Temperature: Ambient
MDL Number: MFCD00016153
CAS Number: 22398-80-7
EINECS: 244-959-5
Merck Index: 13,04976
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty