Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt disulfide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cobalt (IV) sulphide Cobaltous sulphide

14005-06 014005-06
AA14005-06EA 173.99 USD InStock
CAAA14005-06 AA14005-06
Cobalt disulfide ≥99.5% (metals basis excluding Ni) (max. 0.2% Ni)
Cobalt disulphide


Formula: CoS₂
MW: 123.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064735
CAS Number: 12013-10-4
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty