Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium stannate hydrate, Reagent Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Disodium tin trioxide Sodium tin(IV) oxide

014021-22 14021-22 014021-36 14021-36
AA14021-22EA 30.69 USD InStock
CAAA14021-36 AA14021-22 AA14021-36 CAAA14021-22
Sodium stannate hydrate, Reagent Grade
Sodium stannate hydrate
Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.


Formula: Na₂O₃Sn
MW: 212.67 g/mol (anhy)
Melting Pt: 140 °C (dec.)
Density: 4,68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014241
CAS Number: 12058-66-1
EINECS: 235-030-5
Merck Index: 13,08752
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty