Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium metasilicate, anhydrous, granular, tech. –18 Mesh

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium metasilicate

014106.A1 014106-D9 14106-D9 14106-A1
AA14106-A1EA 64.92 USD InStock
CAAA14106-D9 AA14106-A1 CAAA14106-A1 AA-14106-D9
Disodium metasilicate, anhydrous, granular, tech. –18 Mesh
di-Sodium metasilicate
Sodium metasilicate is used as a starting material for zeolites and silica catalysts. It acts as an adhesive and binder, corrosion inhibitor, penetrating sealant, coagulant in waste water treatment, fixative for hand dyeing with reactive dye and detergent auxiliaries.

It is also used in boiler compounds, cleaners, bleaches, aluminum paint strippers and washing carbonated drinking bottles. It finds application in cements, drilling fluids, fireproofing wood, refractories and automobiles. Further, it serves as a surfactant, emulsifier, wetting agent, suspending agent, dispersing agent and anti-corrosive agent in boiler water.

Incompatible with acids, bronze, aluminum, brass, copper, lead, tin, zinc and other alkali sensitive metals.


Formula: Na₂O₃Si
MW: 122.06 g/mol
Melting Pt: 1088 °C
Density: 2.61
Storage Temperature: Ambient
MDL Number: MFCD00003492
CAS Number: 6834-92-0
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 12,08788
Order Now
Specification Test Results
Assay (Unspecified), SiO₂ 44.7 - 47.6%
Assay (Unspecified), Na₂O 49.1 - 51.7%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty