Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) sulfate pentahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Copper (II) sulfateCopper (II) sulfate 5-hydrateCopper sulfate pentahydrateCupric sulphate pentahydrate

014178-A3 14178-36 014178-36 14178-A3
AA14178-A3EA 253.93 USD InStock
AA14178-A3 AA14178-36 CAAA14178-36 CAAA14178-A3
Copper(II) sulfate pentahydrate 98.0-102.0% ACS
Copper (II) sulphate pentahydrate


Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.270
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
REACH: 01-2119520566-40
Merck Index: 13,02682
Order Now
Specification Test Results
Insoluble matter 0.005%
Cl 0.001%
Nitrogen compounds (as N) 0.002%
Ca 0.005%
Ni 0.005%
K 0.01%
Na 0.0002
Fe 0.00003

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty