Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum silicate

Supplier: Thermo Scientific Chemicals
Synonyms: Aluminum hydroxide silicate (2:4:1)Silicic acid (H6Si2O7), Aluminum salt (1:2) dihydrateAluminum hydroxide silicate (Al2(OH)4(Si2O5))Dialuminum trioxido-Trioxidosilyloxy-silane dihydrateKaolinite

14231-A3 14231-36 14231.36 014231.A3
AA14231-36EA 62.76 USD InStock
AA14231-36 AA14231-A3 CAAA14231-36 CAAA14231-A3
Aluminum silicate
Aluminum silicate
Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.


Formula: Al₂Si₂O₇.2H₂O
MW: 258.16 g/mol (222.13 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 12428-46-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty