Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenate decahydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

014239-14 014239-22 14239-22 14239-14
AA14239-14EA 107.56 USD InStock
AA14239-14 AA14239-22 CAAA14239-14 CAAA14239-22
Sodium selenate decahydrate ≥99.9% (metals basis)
Sodium selenate decahydrate


Formula: Na₂SeO₄·10H₂O
MW: 369.09 g/mol
Melting Pt: 35 °C (dec.)
Density: 1,600
Storage Temperature: Ambient
MDL Number: MFCD00149174
CAS Number: 10102-23-5
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty