Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium nitrite ≥96.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

014245-09 014245-22 014245-36
AA14245-22EA 94.97 USD InStock
AA14245-22 AA14245-36 AA14245-09
Potassium nitrite ≥96.0% ACS
Potassium nitrite
In analytical chemistry (testing for amino acids, cobalt, iodine, urea)


Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 350 °C (dec.)
Density: 1.915
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734
Order Now
Specification Test Results
Insoluble matter 0.01%
Cl 0.03%
SO3 Pass test
Ca 0.005%
Mg 0.002%
Heavy Metals (as Pb) 0.001%
Fe 0.001%
Na 0.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty