Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) bromide Cu ≥ 28.1%, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cuprous bromide

14258-36 014258-36 014258-22 14258-22
AA14258-22EA 60.79 USD InStock
AA14258-22 CAAA14258-22 AA14258-36 CAAA14258-36
Copper(II) bromide Cu ≥ 28.1%, REacton®, Reagent Grade
Copper (II) bromide


Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900 °C
Melting Pt: 498 °C
Density: 4.710
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Merck Index: 13,02656
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty