Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cesium nitrate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nitric acid cesium salt

014440-14 014440-36 014440-22 014440-06
AA14440-14EA 104.99 USD InStock
AA14440-14 AA14440-36 AA14440-22 AA14440-06
Cesium nitrate ≥99.99% (metals basis)
Cesium nitrate


Formula: CsNO₃
MW: 194.91 g/mol
Boiling Pt: Dec.
Melting Pt: 414 °C
Density: 3.68
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1,III
Merck Index: 12,02057
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty