Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Manganous chloride Manganese dichloride

14464-06 014464-14 14464-14 014464-06 14464-22 14464-22
AA14464-14EA 273.99 USD InStock
AA14464-14 CAAA14464-06 AA14464-06 CAAA14464-22 AA14464-22 CAAA14464-14
Manganese(II) chloride, ultra dry ≥99.99% (metals basis)
Manganese (II) chloride


Formula: Cl₂Mn
MW: 125.84 g/mol
Boiling Pt: 1190 °C
Melting Pt: 650 °C
Density: 2.977
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05768
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty