Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride, ultra dry ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Potassiumchloride

14466-18 14466-09 14466-30 14466-18 14466-09 14466-30
AA14466-18EA 266 USD InStock
CAAA14466-18 AA14466-18 CAAA14466-09 AA14466-09 CAAA14466-30 AA14466-30
Potassium chloride, ultra dry ≥99.95% (metals basis)
Potassium chloride


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1500 °C (subl.)
Melting Pt: 773 °C
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty